Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bismuth(III) oxide, 99.9%, extra pure, ACROS Organics™
Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
Specifications
Assay Percent Range | 99.9% |
Linear Formula | Bi2O3 |
Fieser | 01,59 |
Merck Index | 15,1273 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in acids |
Formula Weight | 465.95 |
Physical Form | Powder |
Percent Purity | 99.9% |
Grade | Extra Pure |
Chlorine (Cl) | 10ppm max. |
Show More |
Safety and Handling
EINECSNumber : 215-134-7