Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis)

Catalog No. p-7048479
Click to view available options
Quantity:
50 g
250 g

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

It is used as a precursor to the preparation of other compounds of bismuth, chemical reagents. It has specialized uses in optical glass, flame-retardant paper, in glaze formulations as a substitute for lead oxides. It is used as solid electrolyte because of its high thermal expansion property. It is used in radio frequency shielding, compound for catheters and other medical tubing products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point ∼1890°C
Melting Point 817°C
Color Yellow
Physical Form Powder
Quantity 50 g
Merck Index 14,1273
Solubility Information Soluble in acids. Insoluble in water & organic solvents.
Formula Weight 465.96
Percent Purity 99.9995%
Grade Puratronic™
Density 8.9 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) oxide
Show More Show Less
EINECSNumber 215-134-7
RTECSNumber EB2984460
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.