Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) oxide, typically 99.99% (metals basis)

Catalog No. AA1713218
Click to view available options
Quantity:
50 g
250 g

Catalog No. AA1713218

Supplier: Thermo Scientific Chemicals 01713218

Only null left
Add to Cart

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

Used as an adhesive and sealant chemical, flame retardant, pigment, propellant and blowing agent. It is also used as a solvent. Other uses are in batteries, electrical and electronic products, explosive materials, ink, toner, and colorant products, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point ca 1890°C
Melting Point 817°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 465.96
Density 8.9
Chemical Name or Material Bismuth(III) oxide
EINECSNumber 215-134-7
RTECSNumber EB2984460
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.