Alfa Aesar™ Bismuth Indium Tin ingot (Field's metal)

Manufacturer: Alfa Aesar™ 4689536

Quantity 500g

 View more versions of this product

Catalog No. AA4689536

  Check Availability
Add to Cart

    Chemical Identifiers

    Chemical Name or Material Bismuth Indium Tin ingot (Field's metal)

    Specifications

    TSCA TSCA
    Physical Form Ingot
    Avg. Mol. Wt. or Mol. Wt. Range Bi:In:Sn; 32.5:51:16.5 wt%
    Quantity 500g
    Chemical Name or Material Bismuth Indium Tin ingot (Field's metal)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit