Learn More
Alfa Aesar™ Bismuth Indium Tin ingot (Field's metal)
Manufacturer:  Alfa Aesar™ 4689522
Quantity 100g
 View more versions of this product
Catalog No. AA4689522
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
Bismuth Indium Tin ingot (Field's metal)
Specifications
Bi:In:Sn; 32.5:51:16.5 wt%
100g
Ingot
Bismuth Indium Tin ingot (Field's metal)
Safety and Handling
SDS & Safety Documents
TSCA : TSCA