Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Nitrate, 5-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Wide Mouth Amber Glass Bottle
10035-06-0
BiH10N3O14
485.067
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth nitrate pentahydrate,bismuth trinitrate pentahydrate,bismuth nitrate, hydrate 1:5,bismuth iii nitrate pentahydrate,unii-fo2icy167b,fo2icy167b,nitric acid, bismuth 3+ salt, pentahydrate,bismuth iii nitrate pentahydrate,acmc-20alle,bismuthnitratepentahydrate
160911
bismuth;trinitrate;pentahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Bi+3]
10035-06-0
BiH10N3O14
485.067
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth nitrate pentahydrate,bismuth trinitrate pentahydrate,bismuth nitrate, hydrate 1:5,bismuth iii nitrate pentahydrate,unii-fo2icy167b,fo2icy167b,nitric acid, bismuth 3+ salt, pentahydrate,bismuth iii nitrate pentahydrate,acmc-20alle,bismuthnitratepentahydrate
160911
bismuth;trinitrate;pentahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Bi+3]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 485.07
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.83kg
Quantity 500g
Chemical Name or Material Bismuth Nitrate, 5-Hydrate