Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Nitrate, Pentahydrate, Crystal, ACS, 98%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10035-06-0
10035-06-0
CAS Max % 1
Arsenic (As) 1e-05
Percent Purity 0.98
Grade ACS
Calcium (Ca) 5e-05
Chloride 1e-05
Copper (Cu) 2e-05
Insoluble Matter 5e-05
Iron (Fe) 1e-05
Lead (Pb) 2e-05
Packaging Amber Glass Bottle
Potassium (K) 0.0001
Silver (Ag) 1e-05
Sodium (Na) 0.0002
Sulfate 5e-05
Quantity 25 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.