Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bismuth subgallate hydrate

Click to view available options
Quantity:
100g
500g
99-26-3
C7H6BiO6
395.099
MFCD01632692
XXCBNHDMGIZPQF-UHFFFAOYSA-L
bismuth subgallate,wismutgallathydroxid,dermatol,bismuth gallate,dermatol puder,bismuth subgallas,bismutum subgallicum,basic bismuth gallate,gallic acid bismuth basic salt,wismutgallat, basisches
16682999
4-hydroxy-1,3,2$l^{2}-benzodioxabismole-6-carboxylic acid;hydrate
C1=C(C=C(C2=C1O[Bi]O2)O)C(=O)O.O
99-26-3
C7H6BiO6
395.099
MFCD01632692
XXCBNHDMGIZPQF-UHFFFAOYSA-L
bismuth subgallate,wismutgallathydroxid,dermatol,bismuth gallate,dermatol puder,bismuth subgallas,bismutum subgallicum,basic bismuth gallate,gallic acid bismuth basic salt,wismutgallat, basisches
16682999
4-hydroxy-1,3,2$l^{2}-benzodioxabismole-6-carboxylic acid;hydrate
C1=C(C=C(C2=C1O[Bi]O2)O)C(=O)O.O
Quantity 500g
Formula Weight 394.10 (Anhydrous)
Chemical Name or Material Bismuth subgallate hydrate