Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Subgallate, USP, 52-57%, Spectrum™ Chemical
SDP

Catalog No. 18601011
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle

B1116, 99-26-3, C7H5BiO6

Spectrum™ Chemical Bismuth Subgallate, USP is used to treat malodor by deodorizing flatulence and stool odors. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 99-26-3
Molecular Formula C7H5BiO6
Molecular Weight (g/mol) 394.09
MDL Number MFCD00044980
InChI Key JAONZGLTYYUPCT-UHFFFAOYSA-K
IUPAC Name 2,7-dihydroxy-2H-benzo[d]1,3-dioxa-2-bismacyclopentane-5-carboxylic acid
SMILES O[Bi]1OC2=C(O1)C(O)=CC(=C2)C(O)=O

Specifications

Packaging Amber Glass Bottle
CAS Max % 100%
Quantity 2.5 kg
Percent Purity 52 to 57%
Grade USP
Chemical Name or Material Bismuth Subgallate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.