Alfa Aesar™ Bismuth Tin eutectic lump, 99.95% (metals basis)

Manufacturer: Alfa Aesar™ 1248036

Quantity 500g

 View more versions of this product

Catalog No. AA1248036

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00143771
    Chemical Name or Material Bismuth Tin eutectic lump

    Specifications

    Physical Form Lumps
    Melting Point 138°C
    TSCA TSCA
    MDL Number MFCD00143771
    Avg. Mol. Wt. or Mol. Wt. Range Bi:Sn; 58:42 wt%
    Quantity 500g
    Chemical Name or Material Bismuth Tin eutectic lump
    Assay 99.95%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit