Learn More
Bisphenol A, 97+%
Description
Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).
Solubility
Soluble in aqueous alkaline solution, acetic acid, alcohol and acetone. Slightly soluble in carbon tetrachloride. It has poor solubility in water.
Notes
Incompatible with acid anhydrides, acid chlorides, strong bases and strong oxidizing agents.

Specifications
Specifications
Melting Point | 153°C to 158°C |
Boiling Point | 200°C (4 mmHg) |
Flash Point | 227°C (440°F) |
Quantity | 1000 g |
Beilstein | 1107700 |
Merck Index | 14,1297 |
Solubility Information | Soluble in aqueous alkaline solution,acetic acid,alcohol and acetone. Slightly soluble in carbon tetrachloride. It has poor solubility in water. |
Formula Weight | 228.29 |
Percent Purity | ≥97% |
Chemical Name or Material | Bisphenol A |
Safety and Handling
Hazard Category | H317-H318-H335-H360F |
Hazard Statement | GHS H Statement H318-H361-H317-H335 Causes serious eye damage. Suspected of damaging fertility or the unborn child. May cause an allergic skin reaction. May cause respiratory irritation. |
Precautionary Statement | P201-P202-P261-P271-P272-P280g-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c |
DOTInformation | Hazard Class: 9; Packaging Group: III |
EINECSNumber | 201-245-8 |
RTECSNumber | SL6300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.