Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bisphenol A bis(chloroformate), 96%, Thermo Scientific™

Catalog No. AC314580100
Click to view available options
Quantity:
10 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2024-88-6
Molecular Formula C17H14Cl2O4
Molecular Weight (g/mol) 353.20
MDL Number MFCD00060109
InChI Key MMWCQWOKHLEYSP-UHFFFAOYSA-N
Synonym bisphenol a bis chloroformate, 2,2-bis 4-chloroformyloxyphenyl propane, bisphenol a dichloroformate, carbonochloridic acid, 1-methylethylidene di-4,1-phenylene ester, 2,2-bis p-chloroformyloxy phenyl propane, carbonochloridic acid, c,c'-1-methylethylidene di-4,1-phenylene ester, isopropylidenedi-p-phenylene bis chloroformate, bisphenol a bischloroformate, bidd:er0407
PubChem CID 74843
ChEBI CHEBI:34576
IUPAC Name 4-(2-{4-[(chlorocarbonyl)oxy]phenyl}propan-2-yl)phenyl carbonochloridate
SMILES CC(C)(C1=CC=C(OC(Cl)=O)C=C1)C1=CC=C(OC(Cl)=O)C=C1

Specifications

Melting Point 90°C to 95°C
Color Brown-Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96%
Linear Formula (ClCOC6H4)C(CH3)
Quantity 10 g
Formula Weight 353.21
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Bisphenol A bis(chloroformate), 96%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.