Learn More
Bisphenol F acetate propionate, epoxy curative, EC-298, Thermo Scientific Chemicals
Description
Bisphenol F acetate propionate does not impede free radical cure, thermoset adhesives, used as curative for epoxy/(meth) acrylate hybrids, hybrid epoxy/free radical thermosets. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Physical Form | Pale yellow liquid |
Density | 1.13 |
Solubility Information | Soluble in ethanol,ether,chloroform,alkali; slightly soluble in DMSO; insoluble in carbon disulfide. |
Quantity | 25 g |
Formula Weight | 298.34 |
Flash Point | 150°C (302°F) |
Chemical Name or Material | Bisphenol F acetate propionate, Epoxy curative, EC-298 |
Safety and Handling
Hazard Category | H302-H319 |
Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.