Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BisphenolA (2,3-dihydroxypropyl) glycidyl ether, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
25 mg
100 mg
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications
Specifications
Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
CAS | 76002-91-0 |
Physical Form | Neat |
Linear Formula | C21H26O5 |
MDL Number | MFCD03453592 |
Synonym | 2-[4-(2,3-Dihydroxypropyloxy)phenyl]-2-[4-(glycidyloxy)phenyl]propane; BADGE-H2O |
Type | Analytical Standard |
Quantity | 100 mg |
Formula Weight | 358.43 |
Percent Purity | ≥95.0% (HPLC) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction