Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bisphosphoglycerate mutase Polyclonal Antibody
Bisphosphoglycerate mutase Polyclonal Antibody
Supplier: Thermo Scientific PA544868
Description
Bisphosphoglycerate mutase Polyclonal Antibody for Western Blot

Specifications
Bisphosphoglycerate mutase | |
Polyclonal | |
Unconjugated | |
BPGM | |
2,3-bisphosphoglycerate mutase; 2,3-bisphosphoglycerate mutase, erythrocyte; 2,3-bisphosphoglycerate synthase; 2,3-diphosphoglycerate mutase; Ab2-098; AI323730; AL022789; Bisphosphoglycerate mutase; BPG-dependent PGAM; Bpgm; C86192; DPGM; erythrocyte 2,3-bisphosphoglycerate mutase; hypothetical protein LOC533785; testis secretory sperm-binding protein Li 202a; zgc:92230 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
669 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P07738 | |
BPGM | |
Synthetic peptide directed towards the middle region of human BPGM. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction