Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bisphosphoglycerate mutase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553434
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553434 Supplier Invitrogen™ Supplier No. PA553434
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRSYNVTPPP IEESHPYYQE IYNDRRYKVC DVPLDQLPRS ESLKDVLERL LPYWNERIAP EVLRGKTILI SAHGNSSRAL LKHLEGISDE DIINITLPTG VPILLELDEN LRAVGPHQFL GDQEAIQAAI KKVEDQGKVK QA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.

Riboflavin kinase is an essential enzyme that catalyzes the phosphorylation of riboflavin to form flavin mononucleotide, an obligatory step in vitamin B2 utilization and flavin cofactor synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bisphosphoglycerate mutase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BPGM
Gene Accession No. P07738, P15327
Gene Alias 2,3-bisphosphoglycerate mutase; 2,3-bisphosphoglycerate mutase, erythrocyte; 2,3-bisphosphoglycerate synthase; 2,3-diphosphoglycerate mutase; Ab2-098; AI323730; AL022789; Bisphosphoglycerate mutase; BPG-dependent PGAM; Bpgm; C86192; DPGM; erythrocyte 2,3-bisphosphoglycerate mutase; hypothetical protein LOC533785; testis secretory sperm-binding protein Li 202a; zgc:92230
Gene Symbols BPGM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Bisphosphoglycerate mutase. Recombinant protein control fragment (Product #RP-90910).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12183, 296973, 669
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.