Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bit1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582473 Supplier Invitrogen™ Supplier No. PA582473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSKTSKTHTD TESEASILGD SGEYKMILVV RNDLKMGKGK VAAQCSHAAV SAYKQIQRRN PEMLKQWEYC GQPKVVVKAP DEETLIALLA HAKMLGLTVS LIQDAGRTQI APGSQTVLGI GPGPADLIDK VTGHLKL Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 81%.

The protein encoded by this gene is a mitochondrial protein with two putative domains, an N-terminal mitochondrial localization sequence, and a UPF0099 domain. In vitro assays suggest that this protein possesses peptidyl-tRNA hydrolase activity, to release the peptidyl moiety from tRNA, thereby preventing the accumulation of dissociated peptidyl-tRNA that could reduce the efficiency of translation. This protein also plays a role regulating cell survival and death. It promotes survival as part of an integrin-signaling pathway for cells attached to the extracellular matrix characterized by intellectual disability, postnatal microcephaly, progressive cerebellar atrophy, hearing impairment, polyneuropathy, failure to thrive, and organ fibrosis with exocrine pancreas insufficiency. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bit1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ptrh2
Gene Accession No. Q9Y3E5
Gene Alias A230072I16Rik; Bcl-2 inhibitor of transcription; bcl-2 inhibitor of transcription 1; BIT1; CFAP37; CGI-147; cilia and flagella associated protein 37; IMNEPD; OTTHUMP00000205936; peptidyl-tRNA hydrolase 2; peptidyl-tRNA hydrolase 2, mitochondrial; PTH; PTH 2; Pth2; Ptrh2; RGD1306819
Gene Symbols Ptrh2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTRH2. Recombinant protein control fragment (Product #RP-90283).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51651
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.