Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BITE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101510 Supplier Invitrogen™ Supplier No. PA5101510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CEP70.

CEP70 plays a role in the organization of both preexisting and nascent microtubules in interphase cells. During mitosis, it is required for the organization and orientation of the mitotic spindle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BITE
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CEP70
Gene Accession No. Q5PQQ9, Q8NHQ1
Gene Alias 6720484E09Rik; BITE; C030018L16Rik; centrosomal protein 70; centrosomal protein 70kDa; Centrosomal protein of 70 kDa; CEP70; I79_009119; p10-binding protein; testicular tissue protein Li 36
Gene Symbols CEP70
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEP70(Accession Q8NHQ1), corresponding to amino acid residues E243-Q293.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367153, 80321
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.