Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biuret, 97%

Catalog No. p-7043447
Click to view available options
Quantity:
100 g
500 g

CAS: 108-19-0 | C2H5N3O2 | 103.081 g/mol

Biuret is used in protein assay. It is also used as Catalytic agent, Petrochemical additive. It is used in organic synthesis. Biuret (Carbamoylurea) was used as a source of dietary nitrogen in purified diets for steers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biuret is used in protein assay. It is also used as Catalytic agent, Petrochemical additive. It is used in organic synthesis. Biuret (Carbamoylurea) was used as a source of dietary nitrogen in purified diets for steers.

Solubility
Soluble in methanol. Partially soluble in cold water, diethyl ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-19-0
Molecular Formula C2H5N3O2
MDL Number MFCD00007946
InChI Key OHJMTUPIZMNBFR-UHFFFAOYSA-N
Synonym biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
PubChem CID 7913
ChEBI CHEBI:18138
IUPAC Name carbamoylurea

Specifications

Melting Point ∽189°C (decomposition)
Density 1.46
Odor Odorless
Linear Formula H2NCONHCONH2
Quantity 100 g
Beilstein 1703510
Merck Index 14,1309
Solubility Information Soluble in methanol. Partially soluble in cold water,diethyl ether.
SMILES C(=O)(N)NC(=O)N
Molecular Weight (g/mol) 103.081
Formula Weight 103.08
Percent Purity 97%
Chemical Name or Material Biuret
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-559-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.