Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biuret Reagent, For Serum Protein, LabChem™

Catalog No. p-5388367
Click to view available options
Quantity:
500 mL
1 L
4 L

Specifications

Chemical Name or Material Biuret Reagent
Color Blue
Name Note For Serum Protein
CAS Max % 99.24, 0.6, 0.15, 0.01
CAS 6381-59-5, 7758-99-8, 1310-73-2
Linear Formula CuSO4·5H2O
Solubility Information Soluble in water
Formula Weight 249.68
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sulfur compounds; Fume; Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.