Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biuret Reagent TS, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
Chemical Name or Material | Biuret Reagent |
Color | Blue |
Name Note | Test Solution |
CAS | 7732-18-5, 1310-73-2, 6381-59-5, 7758-99-8 |
CAS Min % | 91.0, 6.22, 0.5, 0.13 |
CAS Max % | 95.0, 6.48, 0.52, 0.13 |
Grade | Laboratory |
Packaging | Cubitainer |
pH | >12 |
Quantity | 20 L |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction