Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BK channel Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980468
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0468 Supplier Bioss Supplier No. BS0729R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.

Specifications

Antigen BK channel
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Kcnma1
Gene Accession No. Q12791
Gene Alias 5730414M22Rik; bA205K10.1; big potassium channel alpha subunit; BK channel; BK channel alpha subunit; BKCa; BKCA alpha; BKCA alpha subunit; BKTM; calcium-activated potassium channel alpha subunit; Calcium-activated potassium channel subunit alpha-1; calcium-activated potassium channel subunit alpha-1; uncharacterized protein; Calcium-activated potassium channel, subfamily M subunit alpha-1; hSlo; K(VCA)alpha; KCa1.1; Kcnma; KCNMA1; KCNMA1b; KCNMA1c; Maxi K channel; MaxiK; maxi-K channel HSLO; mSlo; mSlo1; potassium calcium-activated channel subfamily M alpha 1; potassium channel, calcium activated large conductance subfamily M alpha, member 1; potassium large conductance calcium-activated channel, subfamily M, alpha member 1; SAKCA; SLO; Slo homolog; Slo1; Slo-alpha; Slowpoke homolog; stretch-activated Kca channel
Gene Symbols Kcnma1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BK channel.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3778
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.