Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences BL21 (DE3) Chemically Competent Cells
SCI_ED

Catalog No. S000746
Change view
Click to view available options
Quantity:
12 x 50 μL
6 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000746 12 x 50 μL
S000752 6 x 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000746 Supplier G-Biosciences Supplier No. 7862029
Add to Cart
Add to Cart

BL21 (DE3) Chemically competent cells are a widely used E. coli strain.

It is deficient in proteases Lon and OmpT, resistance to phage T1 (fhuA). It is suitable for protein expression under T7 promoter.

Specifications

Product Type Competent Cells
Sterility Sterile
Species E. coli
Format Vial
For Use With (Application) DNA Transformation
Quantity 12 x 50 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.