Learn More
G-Biosciences BL21 (DE3) Electro Competent Cells

Description
It is deficient in proteases Lon and OmpT, resistance to phage T1 (fhuA). It is suitable for protein expression under T7 promoter.
Specifications
Specifications
| Product Type | Competent Cells |
| Sterility | Sterile |
| Species | E. coli |
| Format | Vial |
| For Use With (Application) | DNA Transformation |
| Quantity | 12 x 50 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.