Learn More
Description
This product is a scalemic mixture of (S) and (R) enantiomers in which (S) is predominant. The exact ratio is batch specific and is expressed as an enantiomeric excess (e.e.) on the Certificate of Analysis.
Specifications
Specifications
| Chemical Name or Material | N-(2-Aminophenyl)-2-((1E,3E)-4-(4-(diethylamino)phenyl)buta-1,3-dien-1-yl)-4,5-dihydrothiazole-4-carboxamide |
| Quantity | 5 mg |
| Target | Bioluminescent Substrates |
| Molecular Formula | C24H28N4OS |
| Purity | 0.9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

