Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLA36 Monoclonal Antibody (EBS-T-002)
GREENER_CHOICE

Catalog No. PIMA533326
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533326 Supplier Invitrogen™ Supplier No. MA533326
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BLA36 is a developmentally regulated 36 kDa antigen expressed on the plasma membrane of B lymphocytes, Reed-Sternberg, and mononuclear Hodgkin's cells. It also gives strong staining of B cell lymphomas including follicular center cell lymphomas (large and small cell types), mantle zone lymphomas, and immunoblastic lymphomas.

BLA.36 is a developmentally regulated 36 kDa antigen expressed on the plasma membrane of B lymphocytes, Reed-Sternberg, and mononuclear Hodgkin's cells. The anti-BLA.36 antibody recognizes all four subtypes of Hodgkin's disease and gives strong staining of B cell lymphomas including follicular center cell lymphomas (large and small cell types), mantle zone lymphomas, and immunoblastic lymphomas. Anti-BLA.36 can be used to distinguish Reed-Sternberg cells and some B-cell lymphomas from other malignant cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLA36
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone EBS-T-002
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias BLA36
Host Species Mouse
Immunogen Human BLA36 recombinant protein.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.