Learn More
Supertek™ Black Enamel Hooked Weights
GREENER_CHOICESCI_ED
Catalog No. p-7250666 Shop All Supertek Scientific Products
Click to view available options
Weight (Metric):
10 g
10, 50, 100, 500, 1000 g, 20 g and 200 g
100 g
1000 g
20 g
200 g
50 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy
The steel black enamel coated hooked weights are designed to link together via their hooks. The bottoms of the hooks are recessed and contain a bar, enabling weights to be hung from each other.
  • Non-movable hook on the top
  • Available in a set of 9 in a molded container, which includes 1 each of 10 g, 50 g, 100 g, 500 g, and 1000 g, and 2 each of 20 g and 200 g
  • All weights can also be sold individually
Specifications
Product Type Hook Weight
Includes Set of 9 in molded container- 1 each of 10 g, 50 g, 100 g, 500 g, and 1000 g, and 2 each of 20 g and 200 g
Weight (Metric) 10, 50, 100, 500, 1000 g, 20 g and 200 g
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.