Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lynn Peavey Black Fluorescent Magnetic Powder
SDPSCI_ED

Catalog No. S69013A Shop All Lynn Peavey Company Products
Click to view available options
Weight (English):
1 oz.
16 oz.

Catalog No. S69013A

Supplier: Lynn Peavey 30009

Only null left
Add to Cart
Add to Cart

Helps identify and preserve the integrity of the fingerprint.

  • Fluorescent feature causes dusted fingerprints to glow intensely when exposed to black light
  • Magnetic feature ensures that latent prints will not be damaged or scratched when powder touches print

Specifications

Color Fluorescent Black
For Use With (Application) For identify and preserve the integrity of the fingerprint
Weight (English) 1 oz.
Product Type Magnetic Fingerprint Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.