Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lynn Peavey Black Fluorescent Magnetic Powder
SDPSCI_ED

Catalog No. p-2829767 Shop All Lynn Peavey Company Products
Change view
Click to view available options
Weight (English):
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
S69013B 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S69013B Supplier Lynn Peavey Supplier No. 30008
Only null left
Add to Cart
Add to Cart

Helps identify and preserve the integrity of the fingerprint.

  • Fluorescent feature causes dusted fingerprints to glow intensely when exposed to black light
  • Magnetic feature ensures that latent prints will not be damaged or scratched when powder touches print

Specifications

Color Fluorescent Black
For Use With (Application) For identify and preserve the integrity of the fingerprint
Weight (English) 16 oz.
Product Type Magnetic Fingerprint Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.