Learn More
Fisherbrand™ Black Phenolic Caps
SDP
Catalog No. FB02912055 Shop All Fisherbrand Products
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
Quantity:
12
576
1600
3100
5500
This item is not returnable. View return policy
This item is not returnable. View return policy
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
Unique cone liner design offers superior torque retention and excellent sealing characteristics
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Quantity 12
Closure Color Black
Neck Finish 24-400
Closure Included No
Product Type Lined Cap
Closure Liner Polyethylene Cone
Closure Material Phenolic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.