Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Black phosphorus

Catalog No. 90289620ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
20 mL

Catalog No. 90-289-620ML

Supplier: Sigma Aldrich 90289620ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Black phosphorus, also known as phosphorene, is a stable allotrope of phosphorus with semiconducting properties. It has a good mechanical property and a tunable band structure. BP also has good anisotropic thermal, electrical and optical properties.Owing to the unique properties such as thickness dependent tunable band gap, strong in-plane anisotropy, and high carrier mobility, 2D-black phosphorous has attracted significant attention recently. The applications of 2D-black phosphorous extend from electronics, to optoelectronics, spintronics, thermoelectrics and storage devices.

Specifications

Linear Formula P
Quantity 20 mL
Synonym 2 D Black phosphorus; Phosphorene
Biological Activity Respiratory system
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.