Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Polypropylene Pulp/Vinyl Lined Caps

Catalog No. 02992959
Click to view available options
Packaging:
Bagged
Bulk

Used for food, beverage, medical and chemical applications packed at <120°F. Qorpak™ Black Polypropylene Pulp/Vinyl Lined Caps are ideal for general use.

  • HDPE coated paper liner
  • Used for mild acids, alkalis, solvents, alcohols, oils and aqueous products
  • Not recommended for active hydrocarbons or bleaches
  • Available in smooth and ribbed style
  • Available as bulk/bagged packaging

Specifications

Color Black
Material Polypropylene
No. per Case 144 Cs.
Packaging Bagged
Neck Finish 100-400
For Use With (Equipment) Wide mouth bottle
Closure Liner Pulp/Vinyl
Closure Type Smooth
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.