Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Blackmaxx Vibration Damping Gloves

Catalog No. 19400059
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are coated with pods of lightweight cellular chloroprene, with encapsulated air to cushion and dampen vibration. The lightweight design reduces hand fatigue in the hands, arms, and shoulders.

  • Nylon/cotton knit material ensures stretch and breathability for extreme user comfort
  • Lightweight cellular chloroprene pods on the palm surface
  • Encapsulated air cushions and dampens vibration
  • Minimized hand fatigue when working with pneumatic or impact tools
  • Ideal for use with sanders, chipping hammers, jackhammers, and trimmers

Specifications

Coating Material Chloroprene
Color Black
Hand Specific
Material Cotton, Nylon
Size Medium
EN388 Standard EN388:3243XX
Finger Style Full
For Use With (Application) Power Tool Operation
For Use With (Equipment) Sanders, Chipping Hammers, Jackhammers, Chain Saws, Grinders, and Trimmers
Glove Cuff Style Knit
Hem Color Red
Series VI473
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.