Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Bladder Slides (Adult Normal)- Paraffin
SDP

Catalog No. NBP261249 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261249 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261249 Supplier Novus Biologicals™ Supplier No. NBP261249
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • Tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures.
  • The repository covers a wide variety of species and diseases, including human adult and fetal normal tissues, human diseased and tumor tissues, as well as mouse, rat, and monkey tissues.
  • All the tissues are identified by a board-licensed pathologist.
  • Patient demographic and disease related history data are available.
  • Formalin fixed paraffin embedded (FFPE) tissue section line encompasses a large selection of tissue sections which are suitable for immunohistochemistry and in situ hybridization assays.
  • The tissue sections are approximately 5 μm thick.
  • For those customers who want to perform FFPE DNA and RNA isolation.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Specifications

Quantity 5 Slides
Storage Requirements Store at 4°C
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.