Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Blade Holder Packages for the Cryotome™ FSE Cryostat, Low-Profile Cryotome Blade Holder

Catalog No. a78910032
Click to view available options
Product Type:
Low Profile Blade Holder

Hold blades for the Cryotome™ FSE Cryostat with these packages.

Universal Blade Holder:
  • Accepts standard C- or D-profile knife up to 220mm; 120mm is standard length
  • Use with standard steel knives or with disposable blade holder
  • Use in conjunction with disposable blade holders (Part Nos. 40 or 47)
Low-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder for use with low-profile disposable blades
High-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder will accept either standard or heavy-duty high-profile blades, providing a wide range of tissue sectioning options

Specifications

Product Type Low Profile Blade Holder
For Use With (Equipment) FSE/FE/SME/E
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.