Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab MasterTech PolyBath

Click to view available options
Quantity:
30 mL
Description
- Virtually eliminates background staining
- Just add five drops to waterbath
- A MasterTech Advanced Diagnostics Component
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Colorless |
| Quantity | 30 mL |
| Grade | Histological |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction