Learn More
DR Instruments Blades for Scalpel Handles
SDPSCI_ED
Catalog No. S08121
Click to view available options
Size:
# 10
# 11
# 12
# 20
# 22
For Use With (Equipment):
Scalpel Handle # 3
Scalpel Handle # 4
Razor edge surgical blades for dissection experiments.
  • Disposable Surgical blades designed for scalpel handles #3 and #4, offering effortless installation and removal.
  • These exceptionally sharp surgical blades facilitate the precise cutting of preserved specimens.
  • Crafted from high-quality carbon steel, they are compatible with scalpel handles No. 3 and 4.
Specifications
Blade Material Carbon Steel
Product Type Blade
Size # 20
For Use With (Equipment) Scalpel Handle # 4
Disposable Single-use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.