Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLAME/SLAMF8 Antibody (242), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290657PCP
Description
BLAME/SLAMF8 Monoclonal antibody specifically detects BLAME/SLAMF8 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
BLAME/SLAMF8 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
56833 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
242 | |
Immunohistochemistry-Paraffin | |
B Lymphocyte Activator Macrophage Expressed, BCM-Like Membrane Protein, B-Lymphocyte Activator Macrophage Expressed, CD353, CD353 Antigen, SBBI42, SLAM Family Member 8, SLAMF8 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse BLAME/SLAMF8 (Accession#: NP_083360.2; Met1-Asp231). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction