Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Blank Non-Reflective Aluminum Sign

Catalog No. p-7147274 Shop All Brady Corporation Products
Click to view available options
Color:
White
Yellow

Abrasion-resistant, chemical-resistant, rigid aluminum panel with protected graphics

  • A rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments
  • Non-Reflective Aluminum (B-555) is ideal for indoor and outdoor usage
  • Printed with durable inks and clear-coat protection for fade resistance
  • Withstands wind, rain, sunlight and high temperatures
  • Average outdoor durability 5-8 years
  • Specifications

    Material Aluminum
    Quantity 1 Ea.
    For Use With (Application) Safety Awareness
    Height (English) 10 in.
    Dimensions (L x W) 10 x 14 in.
    Color Yellow

    Adhere to
    Safety Regulations and Best Practices

    Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.