Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLAST-2 (CD23) Mouse anti-Human, Unconjugated, Clone: EBVCS2, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
| Antigen | BLAST-2 (CD23) |
|---|---|
| Clone | EBVCS2 |
| Applications | Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF22712MI
|
MilliporeSigma
MABF227125UG |
25 μg |
Each for $189.29
|
|
|||||
|
MABF22711MI
|
MilliporeSigma
MABF2271100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
BLAST-2 (CD23) Monoclonal antibody specifically detects BLAST-2 (CD23) in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
| BLAST-2 (CD23) | |
| Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| Low affinity immunoglobulin epsilon Fc receptor;C-type lectin domain family 4 member J;CD23;Fc-epsilon-RII;Immunoglobulin E-binding factor;Lymphocyte IgE receptor | |
| EBV-transformed human cells. | |
| Primary | |
| Stable for 1 year at 2 to 8°C from date of receipt. | |
| 36.47 kDa calculated. |
| EBVCS2 | |
| Monoclonal | |
| Purified | |
| Inflammation and Immunology | |
| NP_001193948 | |
| FCER2, CD23A, CLEC4J, FCE2, IGEBF | |
| IgG1 κ | |
| Protein G Purified | |
| Clone EBVCS2 is a mouse monoclonal antibody that detects Low affinity immunoglobulin epsilon Fc receptor (CD23; BLAST-2). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title