Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLAST-2 (CD23) Mouse anti-Human, Unconjugated, Clone: EBVCS3, MilliporeSigma™

Catalog No. MABF23182MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF23182MI 25 μg
MABF23181MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF23182MI Supplier MilliporeSigma Supplier No. MABF231825UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BLAST-2 (CD23) Monoclonal antibody specifically detects BLAST-2 (CD23) in Human samples. It is validated for Activity Assay, Flow Cytometry, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLAST-2 (CD23)
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone EBVCS3
Conjugate Unconjugated
Gene Accession No. NP_001193948.2
Gene Alias Low affinity immunoglobulin epsilon Fc receptor;C-type lectin domain family 4 member J;CD23;Fc-epsilon-RII;Immunoglobulin E-binding factor;Lymphocyte IgE receptor
Gene Symbols FCER2, CD23A, CLEC4J, FCE2, IGEBF
Host Species Mouse
Immunogen EBV-transformed human cells.
Molecular Weight of Antigen 36.47 kDa calculated.
Purification Method Ligatrap
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone EBVCS3 is a mouse monoclonal antibody that detects Low affinity immunoglobulin epsilon Fc receptor (CD23; BLAST-2).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.