Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blasticidin S hydrochloride, 98+%, 10mg/ml in 1M HEPES buffer soln.

Catalog No. p-7057396
Click to view available options
Quantity:
20 mg

Inhibits protein synthesis and induces apoptosis | CAS: 3513-03-9 | C17H27ClN8O5 | 458.90 g/mol

Blasticidin S hydrochloride is used to inhibit protein synthesis in prokaryotic and eukaryotic organisms. It also induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Blasticidin S hydrochloride is used to inhibit protein synthesis in prokaryotic and eukaryotic organisms. It also induces apoptosis.

Solubility
Soluble in water.

Notes
Store in cool place. Recommended to prepare and use solutions on the same day. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3513-03-9
Molecular Formula C17H27ClN8O5
Molecular Weight (g/mol) 458.90
MDL Number MFCD02091640
InChI Key YQXYQOXRCNEATG-UHFFFAOYNA-N
PubChem CID 124080960
SMILES CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl

Specifications

Color Colorless
Physical Form Liquid
Quantity 20 mg
UN Number UN2810
Merck Index 14,1316
Solubility Information Soluble in water.
IUPAC Name (2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride
Formula Weight 458.9
Percent Purity ≥98%
Chemical Name or Material Blasticidin S hydrochloride, 10mg/mL in 1M HEPES buffer soln.
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.