Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BLCAP Monoclonal Antibody (4E1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA532921
Description
BLCAP Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a tumor suppressor protein that reduces cell growth by stimulating apoptosis. The first intron of this gene contains the neuronatin gene, which is imprinted and only expressed from the paternal allele. This gene is imprinted in brain where it is differentially expressed from different promoters. Transcription from the upstream promoter occurs preferentially on the maternal allele, and transcripts are preferentially expressed from the downstream promoter on the paternal allele. Alternative promoters and alternative splicing result in multiple transcript variants that encode the same protein.[provided by RefSeq, Nov 2009].
Specifications
BLCAP | |
Monoclonal | |
2 mg/mL | |
PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4 | |
P62950, P62952 | |
BLCAP | |
Synthetic peptide within Human BLCAP aa 38-87. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
4E1 | |
Unconjugated | |
BLCAP | |
AI462828; Bc10; bladder cancer 10 kDa protein; bladder cancer associated protein; bladder cancer associated protein homolog; bladder cancer associated protein homolog (human); bladder cancer related protein; bladder cancer related protein (10kD); bladder cancer-associated protein; BLCAP; Blcapbladder cancer associated protein homolog | |
Mouse | |
Protein G | |
RUO | |
10904, 171113 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction