Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLCAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565469 Supplier Invitrogen™ Supplier No. PA565469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NCFLYHCSDSP LPESAHDPGV VG Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

This gene encodes a tumor suppressor protein that reduces cell growth by stimulating apoptosis. The first intron of this gene contains the neuronatin gene, which is imprinted and only expressed from the paternal allele. This gene is imprinted in brain where it is differentially expressed from different promoters. Transcription from the upstream promoter occurs preferentially on the maternal allele, and transcripts are preferentially expressed from the downstream promoter on the paternal allele. Alternative promoters and alternative splicing result in multiple transcript variants that encode the same protein.[provided by RefSeq, Nov 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLCAP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BLCAP
Gene Accession No. P62952
Gene Alias AI462828; Bc10; bladder cancer 10 kDa protein; bladder cancer associated protein; bladder cancer associated protein homolog; bladder cancer associated protein homolog (human); bladder cancer related protein; bladder cancer related protein (10kD); bladder cancer-associated protein; BLCAP; Blcapbladder cancer associated protein homolog
Gene Symbols BLCAP
Host Species Rabbit
Immunogen Recombinant Human BLCAP. Recombinant protein control fragment (Product #RP-106146).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10904
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.