Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Bleomycin sulfate
SDP

Catalog No. RD10187998 Shop All Novus Products
Change view
Click to view available options
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
RD10187998 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10187998 Supplier R&D Systems™ Supplier No. 7147/5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Tocris product, part of R&D Systems.

Bleomycin sulfate is a glycopeptide antibiotic produced by Streptomyces verticillus. It inhibits DNA synthesis and causes single and double-strand breaks at specific base sequences in intracellular DNA of bacterial and mammalian cells. It also acts as an inducer and regulator of apoptosis and inhibits tumor angiogenesis. Anticancer agent for squamous cell carcinomas (SCC) with IC50 = 4 nM in UT-SCC-19A cells. In vivo, Bleomycin sulfate induces pulmonary inflammation and fibrosis in lung injury models (mice).

Bleomycin sulfate contains two main components:

1) Bleomycin A2 hemisulfate, C55H84N17O21S3.HSO4, molecular weight = 1512.62 Da

2) Bleomycin B2, C55H84N20O21S2, molecular weight = 1425.52 Da.

The exact proportions vary from batch to batch and can be found on the relevant certificate of analysis.

Specifications

CAS 9041-93-4
Quantity 5 mg
Product Line Tocris
Target DNA, RNA and Protein Synthesis Inhibitors
Molecular Formula C55H84N17O21S3.HSO4 (for A2)
Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.