Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLIMP1/PRDM1 Antibody (3H2-E8), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600235R
Description
BLIMP1/PRDM1 Monoclonal specifically detects BLIMP1/PRDM1 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| BLIMP1/PRDM1 | |
| Monoclonal | |
| DyLight 550 | |
| O75626 | |
| PRDM1 | |
| A fragment of mouse Blimp-1 corresponding to residues 199-409. [UniProt# Q60636] | |
| 0.1 mL | |
| Chromatin Research, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Zinc Finger | |
| 639 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3H2-E8 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PR-domain zinc finger protein 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction