Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLIMP1/PRDM1 Antibody (646702), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB36081B
Description
BLIMP1/PRDM1 Monoclonal specifically detects BLIMP1/PRDM1 in Human samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry.Specifications
BLIMP1/PRDM1 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry | |
646702 | |
Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PR-domain zinc finger protein 1 | |
E. coli-derived recombinant human BLIMP1/PRDM1, Lys667-Cys789, Accession # O75626 | |
0.1 mL | |
B Cell Development and Differentiation Markers, Cancer, Chromatin Research, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Zinc Finger | |
639 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction