Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLK Monoclonal Antibody (9D10B7)
GREENER_CHOICE

Catalog No. PIMA515298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515298 Supplier Invitrogen™ Supplier No. MA515298
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15298 targets BLK in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15298 immunogen is purified recombinant fragment of BLK expressed in E. Coli.

B-lymphoid tyrosine kinase (BLK) is a non-receptor tyrosine kinase that belongs to the Src family of kinases. BLK plays an important role in B-cell activation. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 9D10B7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene BLK
Gene Accession No. P51451
Gene Alias B lymphocyte kinase; B lymphoid kinase; B lymphoid tyrosine kinase; Blk; BLK nonreceptor tyrosine kinase; BLK proto-oncogene, Src family tyrosine kinase; MGC10442; MODY11; p55-Blk; tyrosine-protein kinase Blk
Gene Symbols BLK
Host Species Mouse
Immunogen Purified recombinant fragment of BLK expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 640
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.