Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blk Polyclonal Antibody, Abbomax

Catalog No. 502203535
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3535 Supplier Abbomax Inc Supplier No. 50010904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Raji Cells. Cellular location: Cytoplasmic.

Blk Polyclonal antibody specifically detects Blk in Human samples. It is validated for Western Blot

Specifications

Antigen Blk
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene BLK
Gene Accession No. P51451
Gene Alias B lymphocyte kinase; B lymphoid kinase; B lymphoid tyrosine kinase; Blk; BLK nonreceptor tyrosine kinase; BLK proto-oncogene, Src family tyrosine kinase; MGC10442; MODY11; p55-Blk; tyrosine-protein kinase Blk
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-term of human Blk protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 640
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.