Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    BLMH/Bleomycin Hydrolase Antibody, mFluor Violet 500 SE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299780MFV500
Description
BLMH/Bleomycin Hydrolase Polyclonal antibody specifically detects BLMH/Bleomycin Hydrolase in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| BLMH/Bleomycin Hydrolase | |
| Polyclonal | |
| Western Blot, ELISA, Immunoprecipitation | |
| BHBMHBLM hydrolase, bleomycin hydrolase, EC 3.4.22.40 | |
| Produced in rabbits immunized with purified, recombinant Human BLMH/Bleomycin Hydrolase (Accession#: NP_000377.1; Ser 2-Glu 455). | |
| 0.1 mL | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 642 | |
| Store at 4C in the dark. | |
| IgG | 
| Western Blot, ELISA, Immunoprecipitation | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction